Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029291.3 | NP_001024462.1 | 10 - 11 | 1422 | 72 | |
NM_001029292.3 | NP_001024463.1 | 10 - 11 | 1401 | 72 | ||
NM_001029294.4 | NP_001024465.1 | 10 - 11 | 1616 | 72 | ||
GenBank mRNA | AF083644.1 | - | 10 - 11 | 1401 | 72 | |
AF083651.1 | - | 10 - 11 | 1593 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |