Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029478.5 | NP_001024649.1 | 5 - 6 | 617 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |