Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029536.5 | NP_001024707.1 | 3 - 4 | 1307 | 58 | |
NM_001381134.1 | NP_001366990.1 | 3 - 4 | 1307 | 58 | ||
GenBank mRNA | AF200706.1 | - | 3 - 4 | 1314 | 58 | |
AF200707.1 | - | 3 - 4 | 1312 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |