Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ikke-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372639.1 | NP_001359523.1 | 5 - 6 | 2339 | 59 | |
NM_181898.4 | NP_871627.1 | 6 - 7 | 2407 | 59 | ||
NM_181899.7 | NP_871628.1 | 6 - 7 | 2402 | 59 |
Gene Symbol: R107.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066600.5 | NP_499001.1 | 5 - 6 | 2338 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |