Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001384004.1 | 25 - 26 | 3879 | 66 | ||
NM_001384005.1 | 7 - 8 | 1167 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |