Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027640.5 | NP_001022811.1 | 8 - 9 | 1588 | 105 | |
NM_001381690.1 | NP_001367960.1 | 11 - 12 | 1889 | 105 | ||
NM_001384005.1 | 8 - 9 | 1560 | 105 | |||
NM_001384006.1 | 7 - 8 | 2005 | 105 | |||
NM_064789.5 | NP_497190.1 | 13 - 14 | 2391 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |