Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372637.3 | NP_001360092.1 | 7 - 8 | 1534 | 102 | |
NM_066533.6 | NP_498934.1 | 6 - 7 | 1495 | 102 | ||
GenBank mRNA | AB110823.1 | - | 5 - 6 | 1558 | 102 | |
AB110824.1 | - | 3 - 4 | 1836 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |