Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nac-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027502.4 | NP_001022673.1 | 5 - 6 | 817 | 66 | |
NM_001379885.1 | NP_001367050.1 | 5 - 6 | 814 | 66 | ||
GenBank mRNA | AY090485.1 | - | 5 - 6 | 814 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |