Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381165.1 | NP_001367039.1 | 1 - 2 | 70 | 96 | |
NM_001381168.2 | NP_001366915.1 | 1 - 2 | 156 | 96 | ||
NM_060212.7 | NP_492613.1 | 1 - 2 | 137 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |