Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026921.5 | NP_001022092.1 | 6 - 7 | 2208 | 78 | |
NM_001306658.3 | NP_001293587.1 | 3 - 4 | 1114 | 78 | ||
NM_001356872.3 | NP_001343760.1 | 3 - 4 | 1168 | 78 | ||
GenBank mRNA | AJ130957.1 | - | 2 - 3 | 1019 | 78 | |
Z32673.1 | - | 1300 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |