Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026921.5 | NP_001022092.1 | 7 - 8 | 2297 | 63 | |
NM_001306658.3 | NP_001293587.1 | 4 - 5 | 1203 | 63 | ||
NM_001356872.3 | NP_001343760.1 | 4 - 5 | 1257 | 63 | ||
GenBank mRNA | AJ130957.1 | - | 3 - 4 | 1108 | 63 | |
Z32673.1 | - | 1389 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |