Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383861.1 | 13 - 14 | 2240 | 115 | ||
NM_062856.7 | NP_495257.3 | 13 - 14 | 2280 | 115 | ||
NM_182120.5 | NP_871920.1 | 2 - 3 | 418 | 115 | ||
GenBank mRNA | AY601656.1 | - | 2 - 3 | 397 | 115 | |
AY601657.1 | - | 2 - 3 | 326 | 115 | ||
AY601658.1 | - | 1 - 2 | 245 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |