Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383861.1 | 15 - 16 | 2667 | 110 | ||
NM_062856.7 | NP_495257.3 | 15 - 16 | 2707 | 110 | ||
NM_182120.5 | NP_871920.1 | 4 - 5 | 845 | 110 | ||
GenBank mRNA | AY601656.1 | - | 4 - 5 | 824 | 110 | |
AY601657.1 | - | 4 - 5 | 753 | 110 | ||
AY601658.1 | - | 3 - 4 | 672 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |