Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063871.6 | NP_496272.2 | 6 - 7 | 2300 | 69 | |
NM_182124.7 | NP_871924.2 | 7 - 8 | 2250 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |