Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047333.6 | NP_001040798.1 | 12 - 13 | 1847 | 91 | |
NM_001306543.3 | NP_001293472.1 | 12 - 13 | 1700 | 91 | ||
NM_001383892.1 | 12 - 13 | 1760 | 91 | |||
NM_063199.6 | NP_495600.1 | 12 - 13 | 1782 | 91 | ||
NM_063200.4 | NP_495601.1 | 10 - 11 | 1880 | 91 | ||
NM_182144.6 | NP_871944.1 | 13 - 14 | 2460 | 91 | ||
NM_182145.5 | NP_871945.1 | 10 - 11 | 1953 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |