Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047333.6 | NP_001040798.1 | 6 - 7 | 723 | 96 | |
NM_001306543.3 | NP_001293472.1 | 6 - 7 | 576 | 96 | ||
NM_001383892.1 | 6 - 7 | 636 | 96 | |||
NM_063199.6 | NP_495600.1 | 6 - 7 | 658 | 96 | ||
NM_063200.4 | NP_495601.1 | 4 - 5 | 756 | 96 | ||
NM_182144.6 | NP_871944.1 | 7 - 8 | 1336 | 96 | ||
NM_182145.5 | NP_871945.1 | 4 - 5 | 829 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |