Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027055.5 | NP_001022226.1 | 1 - 2 | 84 | 78 | |
NM_061310.6 | NP_493711.2 | 1 - 2 | 78 | 78 | ||
NM_061311.6 | NP_493712.1 | 1 - 2 | 80 | 78 | ||
NR_132575.1 | 1 - 2 | 76 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |