Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330963.2 | NP_001317775.1 | 7 - 8 | 1717 | 122 | |
NM_001330964.4 | NP_001317776.1 | 7 - 8 | 1723 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |