Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368589.3 | NP_001355398.1 | 15 - 16 | 2202 | 83 | |
NM_001368590.4 | NP_001355414.1 | 5504 | 83 | |||
NM_001381447.1 | NP_001367591.1 | 14 - 15 | 2240 | 83 | ||
NM_001381448.2 | NP_001367590.1 | 14 - 15 | 2251 | 83 | ||
NM_062827.5 | NP_495228.1 | 13 - 14 | 2498 | 83 | ||
GenBank mRNA | AY887905.1 | - | 13 - 14 | 2520 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |