Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368589.3 | NP_001355398.1 | 4 - 5 | 414 | 78 | |
NM_001368590.4 | NP_001355414.1 | 3716 | 78 | |||
NM_001381447.1 | NP_001367591.1 | 3 - 4 | 452 | 78 | ||
NM_001381448.2 | NP_001367590.1 | 3 - 4 | 463 | 78 | ||
NM_062827.5 | NP_495228.1 | 2 - 3 | 710 | 78 | ||
GenBank mRNA | AY887905.1 | - | 2 - 3 | 732 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |