Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372906.2 | NP_001359554.1 | 6 - 7 | 929 | 145 | |
NM_001380412.1 | NP_001368470.1 | 6 - 7 | 917 | 145 | ||
NM_069300.5 | NP_501701.2 | 8 - 9 | 1109 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |