Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lst-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307068.3 | NP_001293997.1 | 1 - 2 | 139 | 95 | |
NM_182290.5 | NP_872090.1 | 4 - 5 | 785 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |