Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029100.4 | NP_001024271.1 | 12 - 13 | 1313 | 120 | |
GenBank mRNA | AB041996.1 | - | 12 - 13 | 1263 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |