Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: marc-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060422.6 | NP_492823.2 | 5 - 6 | 2531 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |