Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sri-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026253.3 | NP_001021424.1 | 5 - 6 | 658 | 106 | |
NM_001026254.4 | NP_001021425.1 | 3 - 4 | 578 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |