Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060496.2 | NP_492897.1 | 2 - 3 | 248 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |