Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060496.2 | NP_492897.1 | 4 - 5 | 533 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |