Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dys-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306310.3 | NP_001293239.1 | 19 - 20 | 4645 | 65 | |
NM_001306311.3 | NP_001293240.1 | 8 - 9 | 1846 | 65 | ||
NM_001306313.4 | NP_001293242.1 | 22 - 23 | 5290 | 65 | ||
NM_001306314.3 | NP_001293243.1 | 18 - 19 | 4612 | 65 | ||
NM_001383731.1 | 18 - 19 | 4663 | 65 | |||
NM_060545.6 | NP_492946.1 | 23 - 24 | 5385 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |