Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313478.3 | NP_001300407.1 | 4 - 5 | 456 | 77 | |
NM_001313479.3 | NP_001300408.1 | 1 - 2 | 162 | 77 | ||
NM_001313480.3 | NP_001300409.1 | 1 - 2 | 162 | 77 | ||
NM_060557.7 | NP_492958.3 | 4 - 5 | 512 | 77 | ||
GenBank mRNA | AY342388.1 | - | 4 - 5 | 456 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |