Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264776.4 | NP_001251705.1 | 4 - 5 | 826 | 77 | |
NM_001373645.1 | NP_001359603.1 | 4 - 5 | 820 | 77 | ||
NM_060575.5 | NP_492976.2 | 4 - 5 | 820 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |