Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060583.4 | NP_492984.1 | 6 - 7 | 1005 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |