Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047239.4 | NP_001040704.1 | 4 - 5 | 1038 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |