Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047239.4 | NP_001040704.1 | 6 - 7 | 1947 | 95 | |
NM_001047240.3 | NP_001040705.1 | 1 - 2 | 534 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |