Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060630.6 | NP_493031.1 | 8 - 9 | 2093 | 60 | |
GenBank mRNA | AY559747.1 | - | 7 - 8 | 1943 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |