Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377649.1 | NP_001364572.1 | 13 - 14 | 3674 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |