Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060749.2 | NP_493150.2 | 3 - 4 | 612 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |