Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060765.2 | NP_493166.1 | 4 - 5 | 1533 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |