Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060857.8 | NP_493258.1 | 1 - 2 | 168 | 62 | |
GenBank mRNA | AY530908.1 | - | 1 - 2 | 125 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |