Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scrm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060920.2 | NP_493321.1 | 3 - 4 | 722 | 70 | |
GenBank mRNA | EF445110.1 | - | 3 - 4 | 703 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |