Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265202.3 | NP_001252131.1 | 12 - 13 | 1624 | 72 | |
NM_001265204.3 | NP_001252133.1 | 2 - 3 | 178 | 72 | ||
GenBank mRNA | AY652944.1 | - | 11 - 12 | 1492 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |