Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265202.3 | NP_001252131.1 | 10 - 11 | 1385 | 75 | |
GenBank mRNA | AY652944.1 | - | 9 - 10 | 1253 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |