Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060965.6 | NP_493366.1 | 6 - 7 | 1637 | 87 | |
GenBank mRNA | AX799789.1 | - | 6 - 7 | 1635 | 87 | |
AY152852.1 | - | 6 - 7 | 1635 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |