Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pry-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061073.6 | NP_493474.2 | 8 - 9 | 1126 | 65 | |
GenBank mRNA | AF468834.1 | - | 8 - 9 | 1128 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |