Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eya-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025884.6 | NP_001021055.1 | 5 - 6 | 644 | 69 | |
NM_001381258.2 | NP_001367120.1 | 5 - 6 | 550 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |