Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zoo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265447.3 | NP_001252376.1 | 11 - 12 | 3013 | 89 | |
NM_001265448.4 | NP_001252377.1 | 10 - 11 | 2916 | 89 | ||
NM_001265449.3 | NP_001252378.1 | 7 - 8 | 2640 | 89 | ||
NM_001377679.2 | NP_001364648.1 | 10 - 11 | 2898 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |