Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026524.7 | NP_001021695.1 | 1 - 2 | 339 | 96 | |
NM_001313528.4 | NP_001300457.1 | 1 | 98 | 96 | ||
NM_001313529.3 | NP_001300458.1 | 1 | 44 | 96 | ||
NM_001313530.3 | NP_001300459.1 | 1 | 44 | 96 | ||
NM_001313531.3 | NP_001300460.1 | 1 | 99 | 96 | ||
NM_001313532.4 | NP_001300461.1 | 1 | 62 | 96 | ||
NM_001381261.2 | NP_001368155.1 | 1 - 2 | 72 | 96 | ||
NM_001381266.2 | NP_001367087.1 | 1 - 2 | 109 | 96 | ||
NM_001383761.1 | 1 - 2 | 179 | 96 | |||
GenBank mRNA | D38540.1 | - | 1 - 2 | 108 | 96 | |
D38541.1 | - | 1 - 2 | 108 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |