Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313524.3 | NP_001300453.1 | 2 - 3 | 295 | 56 | |
NM_001383762.1 | 2 - 3 | 296 | 56 | |||
NM_001383763.1 | 2 - 3 | 296 | 56 | |||
GenBank mRNA | D38542.1 | - | 2 - 3 | 296 | 56 | |
LC215398.1 | - | 2 - 3 | 295 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |