Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026292.5 | NP_001021463.1 | 13 - 14 | 3479 | 104 | |
NM_061174.6 | NP_493575.2 | 13 - 14 | 3474 | 104 | ||
GenBank mRNA | U33051.1 | - | 13 - 14 | 3475 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |