Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026292.5 | NP_001021463.1 | 6 - 7 | 1401 | 59 | |
NM_061174.6 | NP_493575.2 | 6 - 7 | 1402 | 59 | ||
GenBank mRNA | U33051.1 | - | 6 - 7 | 1403 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |