Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061174.6 | NP_493575.2 | 7 - 8 | 1519 | 76 | |
GenBank mRNA | U33051.1 | - | 7 - 8 | 1520 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |